Assay Details
| Gene Symbol: Tdrd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002238.2 | NP_001002238.1 | 4 - 5 | 592 | 88 | |
| NM_001002240.2 | NP_001002240.1 | 4 - 5 | 565 | 88 | ||
| NM_001002241.2 | NP_001002241.1 | 3 - 4 | 865 | 88 | ||
| NM_031387.3 | NP_113564.2 | 4 - 5 | 725 | 88 | ||
| GenBank mRNA | AB067571.1 | - | 3 - 4 | 546 | 88 | |
| AB183526.1 | - | 4 - 5 | 592 | 88 | ||
| AB183527.1 | - | 4 - 5 | 565 | 88 | ||
| AB183528.1 | - | 3 - 4 | 865 | 88 | ||
| AB183529.1 | - | 3 - 4 | 429 | 88 | ||
| AK029806.1 | - | 4 - 5 | 726 | 88 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AB183529.1 transcripts.
Set Membership: |
|

Tdrd1
Genomic Map