Assay Details
| Gene Symbol: Pnliprp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011128.2 | NP_035258.2 | 1 - 2 | 130 | ||
| GenBank mRNA | AK008021.1 | - | 1 - 2 | 130 | ||
| AK131882.1 | - | 1 - 2 | 130 | |||
| BC094923.1 | - | 1 - 2 | 6431510 | 130 | ||
| M30687.1 | - | 1 - 2 | 130 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Pnliprp2
Genomic Map