Assay Details
| Gene Symbol: Pnliprp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011128.2 | NP_035258.2 | 6 - 7 | 802 | 69 | |
| GenBank mRNA | AK131882.1 | - | 6 - 7 | 788 | 69 | |
| BC094923.1 | - | 6 - 7 | 761 | 6431510 | 69 | |
| M30687.1 | - | 6 - 7 | 795 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pnliprp2
Genomic Map