Assay Details
| Gene Symbol: Pnkd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039509.1 | NP_001034598.1 | 6 - 7 | 651 | 63 | |
| NM_019999.2 | NP_064383.1 | 5 - 6 | 560 | 63 | ||
| GenBank mRNA | AB041609.1 | - | 5 - 6 | 561 | 63 | |
| AK041238.1 | - | 5 - 6 | 494 | 63 | ||
| AK173125.1 | - | 6 - 7 | 625 | 63 | ||
| BC058945.1 | - | 7 - 8 | 747 | 6812127 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pnkd
Genomic Map