Assay Details
| Gene Symbol: Eef1b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018796.3 | NP_061266.2 | 5 - 6 | 901 | 126 | |
| GenBank mRNA | AF029844.2 | - | 5 - 6 | 567 | 126 | |
| AK012756.1 | - | 5 - 6 | 604 | 126 | ||
| AK027911.1 | - | 5 - 6 | 604 | 126 | ||
| AK168191.1 | - | 5 - 6 | 603 | 126 | ||
| BC003899.1 | - | 5 - 6 | 597 | 3600850 | 126 | |
| BC023139.1 | - | 5 - 6 | 613 | 3963930 | 126 | |
| BC039635.1 | - | 5 - 6 | 616 | 126 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20285515 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20285516 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20285517 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P412595 | AF029844.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P412595 | AF029844.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 36701 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 315955 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P177185 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P412595 | AF029844.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P412595 | AF029844.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P412595 | AF029844.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P412595 | AF029844.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20285534 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20285541 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20285528 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20285529 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20285538 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20285517 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20285527 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20285526 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20285544 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20285515 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20285519 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20285521 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20285522 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20285537 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20285523 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20285536 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20285524 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P177185 | - | - |
| Whole Mouse Genome G4122A | A_51_P412595 | AF029844.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P412595 | AF029844.2 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P177185 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00488056 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00488054 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20285515 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20285517 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20285516 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20285522 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20285521 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20285536 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20285544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20285523 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20285515 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20285534 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20285543 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20285517 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20285514 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20285541 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20285528 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20285512 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20285510 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20285538 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20285519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20285526 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20285534 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20285517 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20285524 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20285515 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20285536 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20285538 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20285526 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20285519 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20285544 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20285543 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00488082 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20285514 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20285542 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20285512 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20285528 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20285541 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20285523 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20285537 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10346868 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10346872 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10346871 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10346870 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10346869 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1448252_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10346867 | - | - |
| Mouse Genome 430 2.0 Array | 1448252_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1448252_PM_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 55949_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 55949_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0100004180.mm.1 | NM_018796.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100004180.mm.1 | NM_018796.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100004180.mm.1 | TC0100000519.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100004177.mm.1 | NM_018796.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100004177.mm.1 | NM_018796.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100004177.mm.1 | TC0100000519.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100004179.mm.1 | NM_018796.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100004179.mm.1 | NM_018796.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100004179.mm.1 | TC0100000519.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100004178.mm.1 | NM_018796.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100004178.mm.1 | NM_018796.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100004178.mm.1 | TC0100000519.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1448252_PM_a_at | - | - |
| Mouse Expression Set 430 | 1448252_a_at | - | - |
| Murine Genome U74v2 Set | 96624_PM_r_at | - | - |
| Murine Genome U74v2 Set | 96624_r_at | - | - |
| Murine Genome U74v2 Set | 169018_at | - | - |
| Murine Genome U74v2 Set | 169018_PM_at | - | - |
| Murine Genome U74v2 Set | 141174_PM_at | - | - |
| Murine Genome U74v2 Set | 141174_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448252_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448252_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448252_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448252_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 907395 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6130161 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4850400 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6130161 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006130161 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004850400 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2708644 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1229042 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1229042 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2708644 | - | - |
| mouse-6 v1.1 (Illumina) | scl000993.1_5-S | - | - |
| mouse-6 v1.1 (Illumina) | scl055949.3_17-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6130161 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4850400 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl055949.3_17-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl000993.1_5-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6130161 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4850400 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2874853 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1229042 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2708644 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1229042 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2708644 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2874853 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl055949.3_17-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl000993.1_5-S | - | - |

Eef1b2
Genomic Map