Assay Details
| Gene Symbol: Ndufs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001160038.1 | NP_001153510.1 | 5 - 6 | 761 | 67 | |
| NM_001160039.1 | NP_001153511.1 | 5 - 6 | 603 | 67 | ||
| NM_001160040.1 | NP_001153512.1 | 4 - 5 | 657 | 67 | ||
| NM_145518.2 | NP_663493.2 | 4 - 5 | 499 | 67 | ||
| GenBank mRNA | AK034597.1 | - | 4 - 5 | 568 | 67 | |
| AK036926.1 | - | 4 - 5 | 364 | 67 | ||
| AK078590.1 | - | 4 - 5 | 345 | 67 | ||
| AK142711.1 | - | 4 - 5 | 379 | 67 | ||
| AK166604.1 | - | 5 - 6 | 450 | 67 | ||
| AK167705.1 | - | 4 - 5 | 347 | 67 | ||
| AK178076.1 | - | 3 - 4 | 269 | 67 | ||
| AK213867.1 | - | 1 - 2 | 109 | 67 | ||
| BC006660.1 | - | 4 - 5 | 386 | 3501043 | 67 | |
| BC015300.1 | - | 4 - 5 | 319 | 4236882 | 67 | |
| BG808119.1 | - | 4 - 5 | 497 | 67 | ||
| BU708393.1 | - | 5 - 6 | 604 | 67 | ||
| CN715066.1 | - | 4 - 5 | 375 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ndufs1
Genomic Map