Assay Details
| Gene Symbol: Ndufs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001160038.1 | NP_001153510.1 | 6 - 7 | 845 | 108 | |
| NM_001160039.1 | NP_001153511.1 | 6 - 7 | 687 | 108 | ||
| NM_001160040.1 | NP_001153512.1 | 5 - 6 | 741 | 108 | ||
| NM_145518.2 | NP_663493.2 | 5 - 6 | 583 | 108 | ||
| GenBank mRNA | AK034597.1 | - | 5 - 6 | 652 | 108 | |
| AK036926.1 | - | 5 - 6 | 448 | 108 | ||
| AK142711.1 | - | 5 - 6 | 463 | 108 | ||
| AK146814.1 | - | 5 - 6 | 601 | 108 | ||
| AK167705.1 | - | 5 - 6 | 431 | 108 | ||
| AK178076.1 | - | 4 - 5 | 353 | 108 | ||
| AK180817.1 | - | 3 - 4 | 233 | 108 | ||
| BC006660.1 | - | 5 - 6 | 470 | 3501043 | 108 | |
| BC015300.1 | - | 5 - 6 | 403 | 4236882 | 108 | |
| CN715066.1 | - | 5 - 6 | 459 | 108 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ndufs1
Genomic Map