Assay Details
| Gene Symbol: Ndufs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001160038.1 | NP_001153510.1 | 8 - 9 | 1051 | 90 | |
| NM_001160039.1 | NP_001153511.1 | 8 - 9 | 893 | 90 | ||
| NM_001160040.1 | NP_001153512.1 | 7 - 8 | 947 | 90 | ||
| NM_145518.2 | NP_663493.2 | 7 - 8 | 789 | 90 | ||
| GenBank mRNA | AK034597.1 | - | 7 - 8 | 858 | 90 | |
| AK036926.1 | - | 7 - 8 | 654 | 90 | ||
| AK078590.1 | - | 7 - 8 | 635 | 90 | ||
| AK142711.1 | - | 7 - 8 | 669 | 90 | ||
| AK146814.1 | - | 7 - 8 | 807 | 90 | ||
| AK166604.1 | - | 8 - 9 | 740 | 90 | ||
| AK167705.1 | - | 7 - 8 | 637 | 90 | ||
| BC006660.1 | - | 7 - 8 | 676 | 3501043 | 90 | |
| BC015300.1 | - | 7 - 8 | 609 | 4236882 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ndufs1
Genomic Map