Assay Details
| Gene Symbol: Ndufs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001160038.1 | NP_001153510.1 | 9 - 10 | 1236 | 86 | |
| NM_001160039.1 | NP_001153511.1 | 9 - 10 | 1078 | 86 | ||
| NM_001160040.1 | NP_001153512.1 | 8 - 9 | 1132 | 86 | ||
| NM_145518.2 | NP_663493.2 | 8 - 9 | 974 | 86 | ||
| GenBank mRNA | AK034597.1 | - | 8 - 9 | 1043 | 86 | |
| AK036926.1 | - | 8 - 9 | 839 | 86 | ||
| AK078590.1 | - | 8 - 9 | 821 | 86 | ||
| AK142711.1 | - | 8 - 9 | 854 | 86 | ||
| AK146814.1 | - | 8 - 9 | 992 | 86 | ||
| AK166604.1 | - | 9 - 10 | 925 | 86 | ||
| AK167705.1 | - | 8 - 9 | 822 | 86 | ||
| BC006660.1 | - | 8 - 9 | 861 | 3501043 | 86 | |
| BC015300.1 | - | 8 - 9 | 794 | 4236882 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ndufs1
Genomic Map