Assay Details
| Gene Symbol: Ino80d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081436.2 | NP_001074905.1 | 1 - 2 | 62 | 93 | |
| NM_001114609.1 | NP_001108081.1 | 1 - 2 | 62 | 93 | ||
| GenBank mRNA | AK049468.1 | - | 1 - 2 | 50 | 93 | |
| BC010584.1 | - | 1 - 2 | 130 | 93 | ||
| BY278053.1 | - | 1 - 2 | 62 | 93 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK049468.1, NM_001081436.2, NM_001114609.1 transcripts.
Set Membership: |
|

Ino80d
Genomic Map