Assay Details
| Gene Symbol: Ino80d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081436.2 | NP_001074905.1 | 6 - 7 | 1246 | 116 | |
| NM_001114609.1 | NP_001108081.1 | 6 - 7 | 1337 | 116 | ||
| GenBank mRNA | AK049468.1 | - | 6 - 7 | 1234 | 116 | |
| AK133321.1 | - | 980 | 116 | |||
| BC010584.1 | - | 6 - 7 | 1405 | 116 | ||
| BC080705.1 | - | 2 - 3 | 776 | 116 | ||
| BC099533.1 | - | 2 - 3 | 764 | 116 | ||
| BC150989.1 | - | 2 - 3 | 782 | 9087902 | 116 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ino80d
Genomic Map