Assay Details
| Gene Symbol: Ino80d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081436.2 | NP_001074905.1 | 9 - 10 | 1717 | 80 | |
| NM_001114609.1 | NP_001108081.1 | 9 - 10 | 1808 | 80 | ||
| GenBank mRNA | AK049468.1 | - | 9 - 10 | 1705 | 80 | |
| AK133321.1 | - | 1451 | 80 | |||
| BC010584.1 | - | 9 - 10 | 1876 | 80 | ||
| BC080705.1 | - | 5 - 6 | 1247 | 80 | ||
| BC099533.1 | - | 5 - 6 | 1235 | 80 | ||
| BC150989.1 | - | 5 - 6 | 1253 | 9087902 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ino80d
Genomic Map