Assay Details
| Gene Symbol: Osgepl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285839.1 | NP_001272768.1 | 6 - 7 | 1418 | 88 | |
| NR_104361.1 | 7 - 8 | 1498 | 88 | |||
| GenBank mRNA | AK011265.1 | - | 6 - 7 | 1366 | 88 | |
| AK045669.1 | - | 6 - 7 | 1380 | 88 | ||
| AK137332.1 | - | 7 - 8 | 1449 | 88 | ||
| BC038910.1 | - | 4 - 5 | 744 | 88 | ||
| BC051211.1 | - | 3 - 4 | 794 | 88 | ||
| BC058172.1 | - | 6 - 7 | 1381 | 5012054 | 88 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Osgepl1
Genomic Map