Assay Details
| Gene Symbol: Arntl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289679.1 | NP_001276608.1 | 8 - 9 | 979 | 119 | |
| NM_001289680.1 | NP_001276609.1 | 8 - 9 | 979 | 119 | ||
| NM_001289681.1 | NP_001276610.1 | 8 - 9 | 903 | 119 | ||
| NM_172309.2 | NP_758513.1 | 8 - 9 | 979 | 119 | ||
| GenBank mRNA | AY005163.1 | - | 8 - 9 | 979 | 119 | |
| AY014836.1 | - | 8 - 9 | 903 | 119 | ||
| BC108965.2 | - | 8 - 9 | 979 | 40047159 | 119 | |
| BC108966.1 | - | 8 - 9 | 979 | 119 | ||
| DQ355143.1 | - | 8 - 9 | 979 | 119 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Arntl2
Genomic Map