Assay Details
| Gene Symbol: Slc40a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016917.2 | NP_058613.2 | 6 - 7 | 1095 | 81 | |
| GenBank mRNA | AF215637.1 | - | 6 - 7 | 1048 | 81 | |
| AF226613.1 | - | 6 - 7 | 1061 | 81 | ||
| AF231120.1 | - | 6 - 7 | 1034 | 81 | ||
| AK008700.1 | - | 6 - 7 | 899 | 81 | ||
| AK032732.1 | - | 1 - 2 | 89 | 81 | ||
| AK144780.1 | - | 6 - 7 | 1082 | 81 | ||
| AK147137.1 | - | 6 - 7 | 1081 | 81 | ||
| AK159294.1 | - | 6 - 7 | 1084 | 81 | ||
| AK159855.1 | - | 6 - 7 | 1081 | 81 | ||
| AK167608.1 | - | 6 - 7 | 1084 | 81 | ||
| BC003438.1 | - | 6 - 7 | 1064 | 2647365 | 81 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK032732.1 transcripts.
Set Membership: |
|

Slc40a1
Genomic Map