Assay Details
| Gene Symbol: Igbp1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015777.2 | NP_056592.2 | 1 | 1220 | 163 | |
| GenBank mRNA | AJ010637.1 | - | 1 | 1220 | 163 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P413134 | BC131968.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P413134 | NM_015777.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P413134 | AJ010637.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P413134 | BC125601.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P413134 | BC131968.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P413134 | NM_015777.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P413134 | AJ010637.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P413134 | BC125601.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 743503 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P141408 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P154178 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P413134 | BC131968.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P413134 | NM_015777.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P413134 | AJ010637.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P413134 | BC125601.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P413134 | BC131968.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P413134 | NM_015777.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P413134 | AJ010637.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P413134 | BC125601.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24885270 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24885236 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24885265 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24885281 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24885260 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24885258 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24885237 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24885254 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24885234 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24885283 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24885279 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24885280 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24885262 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24885263 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24885247 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24885266 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24885249 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24885241 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24885277 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24885248 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24885268 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24885251 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24885274 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24885245 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24885238 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24885240 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24885257 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24885264 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24885252 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24885253 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24885244 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24885255 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24885275 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24885276 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P154178 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P141408 | - | - |
| Whole Mouse Genome G4122A | A_52_P413134 | BC131968.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P413134 | NM_015777.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P413134 | AJ010637.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P413134 | BC125601.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P413134 | BC131968.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P413134 | NM_015777.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P413134 | AJ010637.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P413134 | BC125601.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P122743 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P141408 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P154178 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05036000 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05275157 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05275151 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24885258 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24885241 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24885256 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24885236 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24885254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24885277 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24885281 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24885231 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24885301 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24885263 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24885238 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24885290 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24885266 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24885251 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24885240 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24885291 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24885247 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24885244 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24885253 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24885233 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24885237 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24885222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24885219 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24885274 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24885280 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24885300 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24885264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24885228 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24885299 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24885302 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24885279 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24885216 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24885230 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24885249 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24885297 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24885224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24885218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24885265 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24885276 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24885294 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24885260 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24885268 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24885262 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24885234 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05275158 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05491743 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05036000 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24885233 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05151921 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24885241 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24885272 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24885275 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05275173 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24885286 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05275157 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24885258 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24885230 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24885231 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24885256 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24885288 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05275160 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24885287 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24885247 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24885270 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24885238 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24885268 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24885236 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05275149 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05809264 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24885283 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24885262 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24885263 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24885274 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24885276 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24885252 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05491739 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24885226 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05275166 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24885260 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24885228 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24885257 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24885248 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24885253 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24885281 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10548949 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548950 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1430512_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10548948 | - | - |
| Mouse Genome 430 2.0 Array | 1430512_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1430512_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 50540_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 50540_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0600026330.mm.1 | TC0600003347.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600026330.mm.1 | NM_015777.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600026330.mm.1 | NM_015777.2 | Refseq |
| Mouse Expression Set 430 | 1430512_PM_a_at | - | - |
| Mouse Expression Set 430 | 1430512_a_at | - | - |
| Murine Genome U74v2 Set | 100287_at | - | - |
| Murine Genome U74v2 Set | 100287_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1430512_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1430512_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1430512_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1430512_PM_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 379501 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6420605 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6420605 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006420605 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2773598 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2773598 | - | - |
| mouse-6 v1.1 (Illumina) | scl28260.1.1_132-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6420605 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl28260.1.1_132-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6420605 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2773598 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2773598 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl28260.1.1_132-S | - | - |

Igbp1b
Genomic Map