Assay Details
| Gene Symbol: Uba2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016682.2 | NP_057891.1 | 13 - 14 | 1411 | 105 | |
| GenBank mRNA | AK075938.1 | - | 4 - 5 | 394 | 105 | |
| AK146925.1 | - | 13 - 14 | 1457 | 105 | ||
| AK151765.1 | - | 13 - 14 | 1416 | 105 | ||
| AK152415.1 | - | 13 - 14 | 1435 | 105 | ||
| AK152977.1 | - | 13 - 14 | 1417 | 105 | ||
| AK163451.1 | - | 13 - 14 | 1407 | 105 | ||
| AK164826.1 | - | 13 - 14 | 1427 | 105 | ||
| AK165321.1 | - | 13 - 14 | 1434 | 105 | ||
| AK166133.1 | - | 13 - 14 | 1443 | 105 | ||
| AK168673.1 | - | 13 - 14 | 1423 | 105 | ||
| AK169168.1 | - | 13 - 14 | 1407 | 105 | ||
| AK180333.1 | - | 1 - 2 | 73 | 105 | ||
| AK212408.1 | - | 1 - 2 | 42 | 105 | ||
| AK213535.1 | - | 1 - 2 | 42 | 105 | ||
| BC054768.1 | - | 13 - 14 | 1437 | 6415071 | 105 | |
| U35833.1 | - | 13 - 14 | 1412 | 105 | ||
| Gene Symbol: Lamb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK196708.1 | - | 125 | 105 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Uba2
Genomic Map