Assay Details
| Gene Symbol: Uba2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016682.2 | NP_057891.1 | 15 - 16 | 1621 | 86 | |
| GenBank mRNA | AK075938.1 | - | 6 - 7 | 604 | 86 | |
| AK146925.1 | - | 15 - 16 | 1667 | 86 | ||
| AK152415.1 | - | 15 - 16 | 1645 | 86 | ||
| AK163451.1 | - | 15 - 16 | 1617 | 86 | ||
| AK164826.1 | - | 15 - 16 | 1637 | 86 | ||
| AK165321.1 | - | 15 - 16 | 1645 | 86 | ||
| AK166133.1 | - | 15 - 16 | 1653 | 86 | ||
| AK168673.1 | - | 15 - 16 | 1633 | 86 | ||
| AK169168.1 | - | 15 - 16 | 1617 | 86 | ||
| AK177509.1 | - | 1 - 2 | 63 | 86 | ||
| AK185145.1 | - | 1 - 2 | 63 | 86 | ||
| AK189273.1 | - | 1 - 2 | 63 | 86 | ||
| AK199297.1 | - | 1 - 2 | 64 | 86 | ||
| AK212310.1 | - | 1 - 2 | 63 | 86 | ||
| AK214571.1 | - | 1 - 2 | 63 | 86 | ||
| BC054768.1 | - | 15 - 16 | 1647 | 6415071 | 86 | |
| U35833.1 | - | 15 - 16 | 1622 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Uba2
Genomic Map