Assay Details
| Gene Symbol: Uba2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016682.2 | NP_057891.1 | 8 - 9 | 790 | 95 | |
| GenBank mRNA | AK042170.1 | - | 1 - 2 | 115 | 95 | |
| AK146925.1 | - | 8 - 9 | 836 | 95 | ||
| AK151765.1 | - | 8 - 9 | 795 | 95 | ||
| AK152415.1 | - | 8 - 9 | 814 | 95 | ||
| AK152977.1 | - | 8 - 9 | 795 | 95 | ||
| AK163451.1 | - | 8 - 9 | 786 | 95 | ||
| AK164826.1 | - | 8 - 9 | 806 | 95 | ||
| AK165321.1 | - | 8 - 9 | 812 | 95 | ||
| AK166133.1 | - | 8 - 9 | 822 | 95 | ||
| AK168673.1 | - | 8 - 9 | 802 | 95 | ||
| AK169168.1 | - | 8 - 9 | 786 | 95 | ||
| AK203168.1 | - | 3 - 4 | 266 | 95 | ||
| AK212719.1 | - | 1 - 2 | 111 | 95 | ||
| AK219873.1 | - | 2 - 3 | 168 | 95 | ||
| BC054768.1 | - | 8 - 9 | 816 | 6415071 | 95 | |
| U35833.1 | - | 8 - 9 | 791 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Uba2
Genomic Map