Assay Details
| Gene Symbol: Uba2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016682.2 | NP_057891.1 | 9 - 10 | 886 | 83 | |
| GenBank mRNA | AK042170.1 | - | 2 - 3 | 211 | 83 | |
| AK146925.1 | - | 9 - 10 | 932 | 83 | ||
| AK151765.1 | - | 9 - 10 | 891 | 83 | ||
| AK152415.1 | - | 9 - 10 | 910 | 83 | ||
| AK163451.1 | - | 9 - 10 | 882 | 83 | ||
| AK164826.1 | - | 9 - 10 | 902 | 83 | ||
| AK165321.1 | - | 9 - 10 | 908 | 83 | ||
| AK166133.1 | - | 9 - 10 | 918 | 83 | ||
| AK168673.1 | - | 9 - 10 | 898 | 83 | ||
| AK169168.1 | - | 9 - 10 | 882 | 83 | ||
| AK212719.1 | - | 2 - 3 | 207 | 83 | ||
| AK219873.1 | - | 3 - 4 | 264 | 83 | ||
| BC054768.1 | - | 9 - 10 | 912 | 6415071 | 83 | |
| U35833.1 | - | 9 - 10 | 887 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Uba2
Genomic Map