Assay Details
| Gene Symbol: Bmpr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007561.4 | NP_031587.1 | 7 - 8 | 1341 | 64 | |
| GenBank mRNA | AF003942.1 | - | 7 - 8 | 1341 | 64 | |
| AK148180.1 | - | 2 - 3 | 159 | 64 | ||
| AK149385.1 | - | 2222 | 64 | |||
| U78048.1 | - | 7 - 8 | 983 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Bmpr2
Genomic Map