Assay Details
| Gene Symbol: Bmpr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007561.4 | NP_031587.1 | 9 - 10 | 1650 | 78 | |
| GenBank mRNA | AF003942.1 | - | 9 - 10 | 1650 | 78 | |
| AK148180.1 | - | 4 - 5 | 468 | 78 | ||
| AK149385.1 | - | 2531 | 78 | |||
| U78048.1 | - | 9 - 10 | 1292 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Bmpr2
Genomic Map