Assay Details
| Gene Symbol: Raph1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045513.3 | NP_001038978.3 | 1 - 2 | 196 | 159 | |
| GenBank mRNA | BI134311.1 | - | 1 - 2 | 196 | 159 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001045513.3 transcripts.
Set Membership: |
|

Raph1
Genomic Map