Assay Details
| Gene Symbol: Raph1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045513.3 | NP_001038978.3 | 7 - 8 | 1295 | 113 | |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Raph1| Gene Symbol: Raph1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045513.3 | NP_001038978.3 | 7 - 8 | 1295 | 113 | |
Set Membership: |
|