Assay Details
| Gene Symbol: Abi2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198127.2 | NP_937760.1 | 3 - 4 | 649 | 115 | |
| GenBank mRNA | BC056345.1 | - | 3 - 4 | 470 | 5705048 | 115 |
More Information
Updated Mapping Information
Set Membership: |
|

Abi2
Genomic Map