Assay Details
| Gene Symbol: Cyp20a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030013.2 | NP_084289.1 | 7 - 8 | 802 | 71 | |
| GenBank mRNA | AK020848.1 | - | 7 - 8 | 853 | 71 | |
| AK053388.1 | - | 7 - 8 | 805 | 71 | ||
| BC049147.1 | - | 6 - 7 | 619 | 6306072 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cyp20a1
Genomic Map