Assay Details
| Gene Symbol: Cyp20a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030013.2 | NP_084289.1 | 9 - 10 | 984 | 83 | |
| GenBank mRNA | AK053388.1 | - | 9 - 10 | 987 | 83 | |
| BC049147.1 | - | 8 - 9 | 801 | 6306072 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cyp20a1
Genomic Map