Assay Details
| Gene Symbol: Nbeal1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173444.2 | NP_775620.2 | 28 - 29 | 4788 | 67 | |
| GenBank mRNA | AK143706.1 | - | 16 - 17 | 2697 | 67 | |
| BC060141.1 | - | 2 - 3 | 653 | 67 | ||
More Information
Set Membership: |
|

Nbeal1
Genomic Map