Assay Details
| Gene Symbol: Nbeal1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173444.2 | NP_775620.2 | 31 - 32 | 5379 | 159 | |
| GenBank mRNA | AK143706.1 | - | 19 - 20 | 3288 | 160 | |
| BC060141.1 | - | 5 - 6 | 1244 | 159 | ||
More Information
Set Membership: |
|

Nbeal1
Genomic Map