Assay Details
| Gene Symbol: Plcz1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_054066.4 | NP_473407.2 | 5 - 6 | 780 | 56 | |
| GenBank mRNA | AF435950.1 | - | 5 - 6 | 785 | 56 | |
| AK006672.1 | - | 5 - 6 | 825 | 56 | ||
| BC106767.1 | - | 5 - 6 | 775 | 40039566 | 56 | |
| BC106768.1 | - | 5 - 6 | 778 | 40039567 | 56 | |
More Information
Updated Mapping Information
Set Membership: |
|

Plcz1
Genomic Map