Assay Details
| Gene Symbol: Bhlhe41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271768.1 | NP_001258697.1 | 1 - 2 | 209 | 86 | |
| NM_024469.2 | NP_077789.1 | 1 - 2 | 209 | 86 | ||
| GenBank mRNA | AA013582.1 | - | 1 - 2 | 210 | 86 | |
| AB044090.1 | - | 1 - 2 | 129 | 86 | ||
| BC023231.1 | - | 1 - 2 | 161 | 86 | ||
| BC037742.1 | - | 1 - 2 | 169 | 86 | ||
| BC051555.1 | - | 1 - 2 | 170 | 86 | ||
| BE985179.1 | - | 4 - 5 | 86 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Bhlhe41
Genomic Map