Assay Details
| Gene Symbol: Bhlhe41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024469.2 | NP_077789.1 | 4 - 5 | 501 | 66 | |
| GenBank mRNA | AB044090.1 | - | 4 - 5 | 421 | 66 | |
| BC023231.1 | - | 4 - 5 | 453 | 66 | ||
| BC037742.1 | - | 4 - 5 | 461 | 66 | ||
| BC051555.1 | - | 4 - 5 | 462 | 66 | ||
| BE985179.1 | - | 1 - 2 | 66 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Bhlhe41
Genomic Map