Assay Details
| Gene Symbol: Dennd5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177192.3 | NP_796166.2 | 8 - 9 | 2477 | 84 | |
| GenBank mRNA | AK038858.1 | - | 8 - 9 | 2378 | 84 | |
| AK050728.1 | - | 8 - 9 | 2335 | 84 | ||
| AK135015.1 | - | 8 - 9 | 2362 | 84 | ||
| BC132199.1 | - | 8 - 9 | 2182 | 40130476 | 84 | |
| BC132535.1 | - | 8 - 9 | 2182 | 40130812 | 84 | |
| BC145128.1 | - | 8 - 9 | 2182 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dennd5b
Genomic Map