Assay Details
| Gene Symbol: Ipo8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081113.1 | NP_001074582.1 | 13 - 14 | 1652 | 121 | |
| GenBank mRNA | BC151036.1 | - | 13 - 14 | 1520 | 9087949 | 121 |
| BC151052.1 | - | 13 - 14 | 1520 | 9087965 | 121 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ipo8
Genomic Map