Assay Details
| Gene Symbol: Ipo8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081113.1 | NP_001074582.1 | 18 - 19 | 2295 | 104 | |
| GenBank mRNA | BC027360.1 | - | 1 - 2 | 117 | 104 | |
| BC034091.1 | - | 5 - 6 | 587 | 104 | ||
| BC151036.1 | - | 18 - 19 | 2163 | 9087949 | 104 | |
| BC151052.1 | - | 18 - 19 | 2163 | 9087965 | 104 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ipo8
Genomic Map