Assay Details
| Gene Symbol: Ipo8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081113.1 | NP_001074582.1 | 2 - 3 | 390 | 91 | |
| GenBank mRNA | AK141137.1 | - | 1 - 2 | 67 | 91 | |
| BC044819.1 | - | 2 - 3 | 349 | 91 | ||
| BC054373.1 | - | 2 - 3 | 362 | 91 | ||
| BC151036.1 | - | 2 - 3 | 258 | 9087949 | 91 | |
| BC151052.1 | - | 2 - 3 | 258 | 9087965 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ipo8
Genomic Map