Assay Details
| Gene Symbol: Ipo8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081113.1 | NP_001074582.1 | 6 - 7 | 951 | 70 | |
| GenBank mRNA | AK047854.1 | - | 4 - 5 | 521 | 70 | |
| AK141137.1 | - | 5 - 6 | 628 | 70 | ||
| BC044819.1 | - | 6 - 7 | 910 | 70 | ||
| BC054373.1 | - | 6 - 7 | 923 | 70 | ||
| BC151036.1 | - | 6 - 7 | 819 | 9087949 | 70 | |
| BC151052.1 | - | 6 - 7 | 819 | 9087965 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ipo8
Genomic Map