Assay Details
| Gene Symbol: Ipo8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081113.1 | NP_001074582.1 | 7 - 8 | 1044 | 110 | |
| GenBank mRNA | AK047854.1 | - | 5 - 6 | 614 | 110 | |
| AK141137.1 | - | 6 - 7 | 721 | 110 | ||
| BC044819.1 | - | 7 - 8 | 1003 | 110 | ||
| BC054373.1 | - | 7 - 8 | 1016 | 110 | ||
| BC151036.1 | - | 7 - 8 | 912 | 9087949 | 110 | |
| BC151052.1 | - | 7 - 8 | 912 | 9087965 | 110 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ipo8
Genomic Map