Assay Details
| Gene Symbol: Tm7sf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026281.2 | NP_080557.2 | 1 - 2 | 209 | 80 | |
| GenBank mRNA | AK010720.1 | - | 1 - 2 | 52 | 80 | |
| AK154633.1 | - | 1 - 2 | 209 | 80 | ||
| AK171111.1 | - | 1 - 2 | 186 | 80 | ||
| AK204708.1 | - | 1 - 2 | 38 | 80 | ||
| BC023389.1 | - | 1 - 2 | 147 | 80 | ||
| BC049960.1 | - | 1 - 2 | 117 | 80 | ||
| DQ104329.1 | - | 1 - 2 | 157 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tm7sf3
Genomic Map