Assay Details
| Gene Symbol: Tm7sf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK171111.1 | - | 7 - 8 | 1045 | 70 | |
| BC023389.1 | - | 7 - 8 | 1007 | 70 | ||
| BC049960.1 | - | 5 - 6 | 704 | 70 | ||
| BC057900.1 | - | 2 - 3 | 170 | 70 | ||
| DQ104329.1 | - | 7 - 8 | 1016 | 70 | ||
More Information
Set Membership: |
|

Tm7sf3
Genomic Map