Assay Details
| Gene Symbol: Clk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042634.2 | NP_001036099.1 | 3 - 4 | 554 | 111 | |
| GenBank mRNA | AK135765.1 | - | 3 - 4 | 489 | 111 | |
| M38381.1 | - | 3 - 4 | 470 | 111 | ||
| X57186.1 | - | 3 - 4 | 454 | 111 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Clk1
Genomic Map