Assay Details
| Gene Symbol: Nif3l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022988.2 | NP_075364.2 | 4 - 5 | 891 | 68 | |
| GenBank mRNA | AF284439.1 | - | 5 - 6 | 1368 | 68 | |
| AK003978.1 | - | 5 - 6 | 999 | 68 | ||
| AK011670.1 | - | 5 - 6 | 908 | 68 | ||
| AK076788.1 | - | 1004 | 68 | |||
| AK131631.1 | - | 5 - 6 | 1254 | 68 | ||
| AK154815.1 | - | 5 - 6 | 983 | 68 | ||
| AK158616.1 | - | 5 - 6 | 1274 | 68 | ||
| BC065163.1 | - | 4 - 5 | 891 | 6404016 | 68 | |
| BC089475.1 | - | 5 - 6 | 818 | 30504614 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nif3l1
Genomic Map