Assay Details
| Gene Symbol: Aox3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023617.2 | NP_076106.2 | 12 - 13 | 1281 | 80 | |
| GenBank mRNA | AF172276.2 | - | 12 - 13 | 80 | ||
| AK004700.1 | - | 1 - 2 | 77 | 80 | ||
| AK030109.1 | - | 8 - 9 | 821 | 80 | ||
| AK149434.1 | - | 12 - 13 | 1242 | 80 | ||
| AY347569.1 | - | 12 - 13 | 1158 | 80 | ||
| AY347570.1 | - | 12 - 13 | 80 | |||
| BC049853.1 | - | 12 - 13 | 1233 | 80 | ||
| BC138876.1 | - | 12 - 13 | 8861898 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Aox3
Genomic Map