Assay Details
| Gene Symbol: Aox3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023617.2 | NP_076106.2 | 26 - 27 | 3185 | 121 | |
| GenBank mRNA | AF172276.2 | - | 26 - 27 | 3261 | 121 | |
| AK004700.1 | - | 15 - 16 | 1981 | 121 | ||
| AK030109.1 | - | 22 - 23 | 2725 | 121 | ||
| AK149434.1 | - | 26 - 27 | 3147 | 121 | ||
| AY347569.1 | - | 26 - 27 | 3062 | 121 | ||
| AY347570.1 | - | 26 - 27 | 3062 | 121 | ||
| BC049853.1 | - | 26 - 27 | 3137 | 121 | ||
| BC138876.1 | - | 26 - 27 | 3164 | 8861898 | 121 | |
More Information
Updated Mapping Information
Set Membership: |
|

Aox3
Genomic Map