Assay Details
| Gene Symbol: Aox4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023631.2 | NP_076120.2 | 11 - 12 | 1237 | 64 | |
| GenBank mRNA | AK158427.1 | - | 11 - 12 | 1238 | 64 | |
| AK160953.1 | - | 11 - 12 | 1203 | 64 | ||
| BC117975.1 | - | 11 - 12 | 1164 | 40098789 | 64 | |
| BC117976.1 | - | 11 - 12 | 1164 | 40098791 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Aox4
Genomic Map