Assay Details
| Gene Symbol: Fam126b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172513.3 | NP_766101.3 | 1 - 2 | 119 | 91 | |
| GenBank mRNA | AK144602.1 | - | 1 - 2 | 56 | 91 | |
| AK168372.1 | - | 1 - 2 | 109 | 91 | ||
| BC049806.1 | - | 1 - 2 | 56 | 91 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK144602.1, AK168372.1, NM_172513.3 transcripts.
Set Membership: |
|

Fam126b
Genomic Map