Assay Details
| Gene Symbol: Fam126b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172513.3 | NP_766101.3 | 5 - 6 | 581 | 99 | |
| GenBank mRNA | AK052638.1 | - | 5 - 6 | 508 | 99 | |
| AK144602.1 | - | 5 - 6 | 518 | 99 | ||
| AK168372.1 | - | 5 - 6 | 571 | 99 | ||
| BC049806.1 | - | 5 - 6 | 518 | 99 | ||
| BC068302.1 | - | 5 - 6 | 489 | 30431671 | 99 | |
More Information
Set Membership: |
|

Fam126b
Genomic Map