Assay Details
| Gene Symbol: Aox2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008419.2 | NP_001008419.1 | 3 - 4 | 308 | 97 | |
| GenBank mRNA | AY187018.2 | - | 3 - 4 | 308 | 97 | |
| AY665589.1 | - | 3 - 4 | 215 | 97 | ||
| BC150826.1 | - | 3 - 4 | 235 | 9087737 | 97 | |
More Information
Set Membership: |
|

Aox2
Genomic Map