Assay Details
| Gene Symbol: Spats2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164566.1 | NP_001158038.1 | 2 - 3 | 177 | 99 | |
| NM_144882.4 | NP_659131.2 | 4 - 5 | 363 | 99 | ||
| GenBank mRNA | AK012791.1 | - | 4 - 5 | 337 | 99 | |
| AK019292.1 | - | 4 - 5 | 337 | 99 | ||
| AK031266.1 | - | 4 - 5 | 318 | 99 | ||
| AK032169.1 | - | 4 - 5 | 298 | 99 | ||
| BC014764.1 | - | 2 - 3 | 177 | 4217648 | 99 | |
| BC021879.1 | - | 4 - 5 | 299 | 4216816 | 99 | |
| BY069430.1 | - | 2 - 3 | 133 | 99 | ||
| BY739337.1 | - | 2 - 3 | 140 | 99 | ||
| EF410080.1 | - | 403 | 99 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Spats2l
Genomic Map