Assay Details
| Gene Symbol: Spats2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164566.1 | NP_001158038.1 | 6 - 7 | 793 | 85 | |
| NM_144882.4 | NP_659131.2 | 8 - 9 | 979 | 85 | ||
| GenBank mRNA | AK031266.1 | - | 8 - 9 | 934 | 85 | |
| AK032169.1 | - | 8 - 9 | 914 | 85 | ||
| BC014764.1 | - | 6 - 7 | 793 | 4217648 | 85 | |
| BC021879.1 | - | 8 - 9 | 915 | 4216816 | 85 | |
| CK624383.1 | - | 1 - 2 | 86 | 85 | ||
| CN698528.1 | - | 2 - 3 | 330 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Spats2l
Genomic Map